Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diaminotoluene, 98%

Catalog No. AC146600250
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC146600250 25 g Glass bottle
AC146602500 250 g Glass bottle
AC146600010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC146600250 Supplier Thermo Scientific Chemicals Supplier No. 146600250
Add to Cart

CAS: 95-80-7 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-80-7
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00007804
InChI Key VOZKAJLKRJDJLL-UHFFFAOYSA-N
Synonym 2,4-diaminotoluene, 2,4-toluenediamine, developer t, toluene-2,4-diamine, 1,3-benzenediamine, 4-methyl, developer mtd, m-toluenediamine, fourrine m, m-tolylenediamine, benzofur mt
PubChem CID 7261
ChEBI CHEBI:34237
IUPAC Name 4-methylbenzene-1,3-diamine
SMILES CC1=CC=C(N)C=C1N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 97.0°C to 101.0°C
Color Brown to Gray
Boiling Point 283.0°C to 285.0°C
Flash Point 149°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula CH3C6H3(NH2)2
Quantity 25 g
Beilstein 13, 124
Solubility Information Solubility in water: 50g/L (25 c). Other solubilities: soluble in alcohol, ether, chloroform and, benzene
Formula Weight 122.17
Percent Purity 98%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2, 4-Diaminotoluene, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
Toxic if swallowed.
Harmful in contact with skin.
May cause damage to organs through prolonged or repeated expos
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Wear protective gloves/protectiv
EINECSNumber 202-453-1
RTECSNumber XS9625000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.