Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromo-1-fluorobenzene 97.0+%, TCI America™
SDP

Catalog No. D34895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34895G 5 g
D348925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D34895G Supplier TCI America Supplier No. D34895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1435-53-6
Molecular Formula C6H3Br2F
Molecular Weight (g/mol) 253.90
MDL Number MFCD00000283
InChI Key MKHDDTWHDFVYDQ-UHFFFAOYSA-N
Synonym 2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j
PubChem CID 15034
IUPAC Name 2,4-dibromo-1-fluorobenzene
SMILES FC1=CC=C(Br)C=C1Br

Specifications

Boiling Point 105°C
Quantity 5 g
Formula Weight 253.90
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dibromo-1-fluorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.