Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromo-6-[(E)-[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]phenol 93.0+%, TCI America™
SDP

Catalog No. D516050MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D516050MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D516050MG Supplier TCI America Supplier No. D516050MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1420042-07-4
Molecular Formula C29H24Br2N2O

Specifications

Quantity 50 mg
Formula Weight 576.33
Percent Purity ≥93.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dibromo-6-[(E)-[[(1R,2R)-2-(isoindolin-2-yl)-1,2-diphenylethyl]imino]methyl]phenol
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.