Learn More
Medchemexpress LLC 2,4-Dibromophenol-d3 | 1219805-97-6 | 99.5% | 254.92 | 100 MG
SDP
Catalog No. 5000452488
Encompass_Preferred
2,4-Dibromophenol-d3 is the deuterium labeled form of 2,4-Dibromophenol. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52488 Supplier Medchemexpress LLC Supplier No. HYW009490S100MG
2,4-Dibromophenol-d3 is the deuterium labeled form of 2,4-Dibromophenol. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has garnered attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS