Learn More
Medchemexpress LLC 2,4-Dibromophenol-d3 | 1219805-97-6 | 99.5% | 254.92 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW009490S
Encompass_Preferred
2,4-Dibromophenol-d3 is the deuterium labeled 2,4-Dibromophenol. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66723