Learn More
Medchemexpress LLC 2,4-Dibromophenol-d3 | 1219805-97-6 | 99.5% | 254.92 | 50 MG
SDP
Catalog No. 5000453364
Encompass_Preferred
2,4-Dibromophenol-d3 is the deuterium labeled 2,4-Dibromophenol. It is used as a tracer and an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-53364 Supplier Medchemexpress LLC Supplier No. HYW009490S50MG
2,4-Dibromophenol-d3 is the deuterium labeled 2,4-Dibromophenol. It is used as a tracer and an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS