Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dibromopyridine, 97%

Catalog No. AC453700010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC453700010 1 g
AC453700050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC453700010 Supplier Thermo Scientific Chemicals Supplier No. 453700010
Add to Cart

CAS: 58530-53-3 | C5H3Br2N | 236.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58530-53-3
Molecular Formula C5H3Br2N
Molecular Weight (g/mol) 236.89
MDL Number MFCD01859720
InChI Key PCMMSLVJMKQWMQ-UHFFFAOYSA-N
Synonym 2,4-dibromo-pyridine, pyridine, 2,4-dibromo, 2,4-dibrompyridin, pubchem5452, acmc-1av5w, 2,4-dibromopyridine, ksc493e4t
PubChem CID 817104
IUPAC Name 2,4-dibromopyridine
SMILES BrC1=CC=NC(Br)=C1

Specifications

Melting Point 36.0°C to 38.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 236.89
Percent Purity 97%
Chemical Name or Material 2,4-Dibromopyridine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.