Learn More
Medchemexpress LLC 2,4-Dibromothiazole | 4175-77-3 | 98.0% | C3HBr2NS | 1 ML
SDP
Catalog No. 5000434769
Encompass_Preferred
2,4-Dibromothiazole is a biochemical reagent utilized as a biological material or organic compound in life science research. It functions as a building block in the synthesis of Melithiazole C through a highly (E)-selective cross-metathesis. Additionally, it serves as an intermediate in preparing various 2,4-disubstituted thiazole derivatives that possess biologically active properties, such as survival motor neuron (SMN) protein modulators.
  • Intended for life science related research.
  • Can be used in the synthesis of complex molecules like Melithiazole C.
  • Applicable in the preparation of biologically active 2,4-disubstituted thiazole derivatives.
  • Available with purity levels of 98.0% and 99.93% (batch-specific).
  • Offered in solid form and in solution (10 mM in DMSO).
Catalog No. 50-004-34769 Supplier Medchemexpress LLC Supplier No. HY5930410MM/1ML
Add to Cart
Add to Cart
2,4-Dibromothiazole is a biochemical reagent utilized as a biological material or organic compound in life science research. It functions as a building block in the synthesis of Melithiazole C through a highly (E)-selective cross-metathesis. Additionally, it serves as an intermediate in preparing various 2,4-disubstituted thiazole derivatives that possess biologically active properties, such as survival motor neuron (SMN) protein modulators.
  • Intended for life science related research.
  • Can be used in the synthesis of complex molecules like Melithiazole C.
  • Applicable in the preparation of biologically active 2,4-disubstituted thiazole derivatives.
  • Available with purity levels of 98.0% and 99.93% (batch-specific).
  • Offered in solid form and in solution (10 mM in DMSO).