Learn More
Medchemexpress LLC 2,4-Dibromothiazole | 4175-77-3 | 98.0% | C3HBr2NS | 100 G
SDP
Catalog No. 5000434253
Encompass_Preferred
2,4-Dibromothiazole is a biochemical reagent used as a biological material or organic compound for life science-related research. It is applied as a building block in the synthesis of Melithiazole C and as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties, such as survival motor neuron (SMN) protein modulators. This product is for research use only.
  • Used as a biological material or organic compound for life science-related research
  • Applied as a building block in the synthesis of Melithiazole C
  • Intermediate in the preparation of various 2,4-disubstituted thiazole derivatives
  • Possesses biologically active properties, such as survival motor neuron (SMN) protein modulators
  • For research use only
Catalog No. 50-004-34253 Supplier Medchemexpress LLC Supplier No. HY59304100G
2,4-Dibromothiazole is a biochemical reagent used as a biological material or organic compound for life science-related research. It is applied as a building block in the synthesis of Melithiazole C and as an intermediate in the preparation of various 2,4-disubstituted thiazole derivatives with biologically active properties, such as survival motor neuron (SMN) protein modulators. This product is for research use only.
  • Used as a biological material or organic compound for life science-related research
  • Applied as a building block in the synthesis of Melithiazole C
  • Intermediate in the preparation of various 2,4-disubstituted thiazole derivatives
  • Possesses biologically active properties, such as survival motor neuron (SMN) protein modulators
  • For research use only