Learn More
Medchemexpress LLC 2,4-Dibromothiazole | 4175-77-3 | 98.0% | C3HBr2NS | 25 G
SDP
Catalog No. 5000358650
Encompass_Preferred
2,4-Dibromothiazole is a biochemical reagent used as a biological material or organic compound for life science-related research. It serves as a building block in synthesizing Melithiazole C, employing highly (E)-selective cross-metathesis. It is also an intermediate in preparing various 2,4-disubstituted thiazole derivatives with biologically active properties, such as survival motor neuron (SMN) protein modulators.
  • Used as a biological material for life science-related research
  • Functions as an organic compound for life science-related research
  • Serves as a building block in synthesizing Melithiazole C
  • Intermediate in preparing various 2,4-disubstituted thiazole derivatives
  • Exhibits biologically active properties, including SMN protein modulation
Catalog No. 50-003-58650 Supplier Medchemexpress LLC Supplier No. 25GHY59304
Add to Cart
Add to Cart
2,4-Dibromothiazole is a biochemical reagent used as a biological material or organic compound for life science-related research. It serves as a building block in synthesizing Melithiazole C, employing highly (E)-selective cross-metathesis. It is also an intermediate in preparing various 2,4-disubstituted thiazole derivatives with biologically active properties, such as survival motor neuron (SMN) protein modulators.
  • Used as a biological material for life science-related research
  • Functions as an organic compound for life science-related research
  • Serves as a building block in synthesizing Melithiazole C
  • Intermediate in preparing various 2,4-disubstituted thiazole derivatives
  • Exhibits biologically active properties, including SMN protein modulation