Learn More
Medchemexpress LLC 2,4-dichloro-3,5-dimethylphenol | 133-53-9 | MFCD00019981 | 95.4% | 191.06 g/mol | C8H8Cl2O | 25 G
SDP
Catalog No. 5000283468
Encompass_Preferred
Dichloroxylenol is a chlorinated phenolic compound used as a bactericide, preservative, and deodorant in biological and veterinary research. It is supplied as a solid for laboratory use and is applied in studies such as endometritis and cervicitis in Holstein cows. Typical identifiers include molecular formula C8H8Cl2O, molecular weight 191.06 g/mol, and CAS number 133-53-9.
  • Bactericidal and preservative properties for antimicrobial research.
  • Solid form suitable for accurate weighing and formulation.
  • High purity appropriate for research applications.
  • Well-characterized chemical identifiers for record keeping and safety documentation.
  • Storage guidance available for long-term and in-solution stability.
Catalog No. 50-002-83468 Supplier Medchemexpress LLC Supplier No. HYW14079925G
Dichloroxylenol is a chlorinated phenolic compound used as a bactericide, preservative, and deodorant in biological and veterinary research. It is supplied as a solid for laboratory use and is applied in studies such as endometritis and cervicitis in Holstein cows. Typical identifiers include molecular formula C8H8Cl2O, molecular weight 191.06 g/mol, and CAS number 133-53-9.
  • Bactericidal and preservative properties for antimicrobial research.
  • Solid form suitable for accurate weighing and formulation.
  • High purity appropriate for research applications.
  • Well-characterized chemical identifiers for record keeping and safety documentation.
  • Storage guidance available for long-term and in-solution stability.