Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-3-ethyl-6-nitrophenol 98.0+%, TCI America™
SDP

Catalog No. D35105G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35105G 5 g
D351025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35105G Supplier TCI America Supplier No. D35105G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 99817-36-4
Molecular Formula C8H6Cl2NO3
Molecular Weight (g/mol) 235.04
MDL Number MFCD00270764
InChI Key YTVCECQSAPGJBB-UHFFFAOYSA-M
Synonym 2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol
PubChem CID 7020339
IUPAC Name 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate
SMILES CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O

Specifications

Melting Point 48°C
Quantity 5 g
UN Number 2811
Formula Weight 236.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-3-ethyl-6-nitrophenol
EINECSNumber (3)-4208
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.