Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-3-pyridinecarboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. D50761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50761G 1 g
D50765G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50761G Supplier TCI America Supplier No. D50761G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 134031-24-6
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 175.996
MDL Number MFCD07437909
InChI Key GWBHJHIZHNTJLA-UHFFFAOYSA-N
Synonym 2,4-dichloronicotinaldehyde, 2,4-dichloropyridine-3-carboxaldehyde, 2,4-dichloro-3-pyridinecarboxaldehyde, 3-pyridinecarboxaldehyde, 2,4-dichloro, pubchem22521, acmc-1bz1r, dichloronicotinaldehyde, ksc518s4r, 2,4-dichloro-3-formylpyridine, 2,4-dichloro-3-pyridinecarbaldehyde
PubChem CID 15053985
IUPAC Name 2,4-dichloropyridine-3-carbaldehyde
SMILES C1=CN=C(C(=C1Cl)C=O)Cl

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 1 g
Formula Weight 176.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-3-pyridinecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.