Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-4'-nitrobiphenyl Ether 98.0+%, TCI America™
SDP

Catalog No. D39125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D39125G Supplier TCI America Supplier No. D39125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1836-75-5
Molecular Formula C12H7Cl2NO3
Molecular Weight (g/mol) 284.092
MDL Number MFCD00128026
InChI Key XITQUSLLOSKDTB-UHFFFAOYSA-N
Synonym nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen
PubChem CID 15787
IUPAC Name 2,4-dichloro-1-(4-nitrophenoxy)benzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl

Specifications

Melting Point 71°C
Color Yellow
Boiling Point 180°C
Quantity 5 g
UN Number 3077
Formula Weight 284.09
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichloro-4′-nitrobiphenyl Ether
EINECSNumber (3)-0658
RTECSNumber KN8400000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.