Learn More
Medchemexpress LLC 2,4-dichloro-5-(((2-((dimethylamino)methyl)phenyl)methoxy)methyl)pyrimidine hydrochloride | 2289726-31-2 | 99.9% | 375.72 | 50 MG
SDP
Catalog No. 5000392904
Encompass_Preferred
DCPT1061 hydrochloride is a potent inhibitor of histone methyltransferases, specifically targeting PRMT1, PRMT6, and PRMT8 in vitro. It exhibits weak inhibitory effects on other epigenetic enzymes like PRMT3, PRMT4, and PRMT5. This compound has demonstrated antitumor effects, making it a promising agent for cancer research.
  • Strong inhibitory effect on PRMT1, PRMT6, and PRMT8.
  • Minimal inhibition of PRMT3, PRMT4, and PRMT5.
  • Exhibits antitumor properties.
  • Suitable for research applications.
Catalog No. 50-003-92904 Supplier Medchemexpress LLC Supplier No. HY162003A50MG
DCPT1061 hydrochloride is a potent inhibitor of histone methyltransferases, specifically targeting PRMT1, PRMT6, and PRMT8 in vitro. It exhibits weak inhibitory effects on other epigenetic enzymes like PRMT3, PRMT4, and PRMT5. This compound has demonstrated antitumor effects, making it a promising agent for cancer research.
  • Strong inhibitory effect on PRMT1, PRMT6, and PRMT8.
  • Minimal inhibition of PRMT3, PRMT4, and PRMT5.
  • Exhibits antitumor properties.
  • Suitable for research applications.