Learn More
Sigma Aldrich 2,4-Dichloro-5,6,7,8-tetrahydroquinazoline
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- 1-methylindole undergoes au(iii)/tppms-catalyzed benzylation reaction with benzhydryl and benzylic alcohols.reactant for preparation of:
- Pharmaceutically active 2-oxo-1-pyrrolidine analogues
- Non-receptor tyrosine kinase (src kinase) inhibitors
- Pet agents for imaging of protein kinase c (pkc)
- Ynediones as highly reactive michael systems
- Anticancer agents
- Polycyclic derivatives of indoles
- Pet agents for imaging of glycogen synthase kinase-3 (gsk-3)
- Anti-prion disease agents
- Bisindole derivatives with antihyperlipidemic activity
- Pet cancer imaging agents
Specifications
Specifications
| Molecular Formula | C8H8Cl2N2 |
| Linear Formula | C8H8Cl2N2 |
| MDL Number | MFCD12400774 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 203.07 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.