Learn More
Accela Chembio Inc 2 | 4-dichloro-5-fluoroquinazoline | 0.25g | 87611-00-5 | MFCD09954883 | 97+% | Shelf Life: 1080 Days | Light Sensitive/moisture Sensitive/store In Freezer(-20) Upon Receipt
Supplier:  Accela Chembio Inc SY0072740.25G
Encompass_Preferred
2 | 4-dichloro-5-fluoroquinazoline | 0.25g | 87611-00-5 | MFCD09954883 | 97+% | Shelf Life: 1080 Days | Light Sensitive/moisture Sensitive/store In Freezer(-20) Upon Receipt
Catalog No. 50-354-638