Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-methylpyridine, 96%, Thermo Scientific™

Catalog No. AAH6163403
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6163403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6163403 Supplier Thermo Scientific Chemicals Supplier No. H6163403

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,4-Dichloro-5-methylpyridine is used as pharmaceutical intermediates.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in a cool, dark and well-ventilated place. Store away from incompatible materials such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56961-78-5
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD08704813
InChI Key DUPJNQNLAKJWKU-UHFFFAOYSA-N
Synonym 2,4-dichloro-5-picoline, pyridine, 2,4-dichloro-5-methyl, 2,4-dichloro-5-methyl-pyridine, pubchem5464, acmc-209lv3, ksc495i9n, 2,4-dichloro-5-methyl pyridine, 2,4-bis chloranyl-5-methyl-pyridine
PubChem CID 12643718
IUPAC Name 2,4-dichloro-5-methylpyridine
SMILES CC1=CN=C(Cl)C=C1Cl

Specifications

Color Colorless
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 162.02
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2,4-Dichloro-5-methylpyridine
Hazard Category H302-H315-H318-H335
Hazard Statement GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.