Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dichloro-5-methylsulfanyl-pyrimidine

Catalog No. SYV000031G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SYV000031G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SYV000031G Supplier Sigma Aldrich Supplier No. SYV000031G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

4-Fluorophenylacetonitrile is starting reagent in the synthesis of 1-Alkyl-N-[2-ethyl-2-(4-fluorophenyl)butyl]piperidine-4-carboxamide derivatives.4-Fluorophenylacetonitrile undergoes biotransformation to 4-Fluorophenylacetic acid by marine fungi, Aspergillus sydowii Ce19.

Specifications

CAS 7401-98-1
Molecular Formula C5H4Cl2N2S
Linear Formula C5H4Cl2N2S
MDL Number MFCD22199809
Quantity 25 g
Molecular Weight (g/mol) 195.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.