Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-5-nitrophenol, 98%, Thermo Scientific™

Catalog No. AAL1836906
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1836906 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1836906 Supplier Thermo Scientific Chemicals Supplier No. L1836906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 39489-77-5
Molecular Formula C6H3Cl2NO3
Quantity 5g
MDL Number MFCD03093072
UN Number UN2811
Beilstein 1733418
Synonym phenol, 2,4-dichloro-5-nitro, 2,4-dichloro-5-nitrobenzenol, 2,4-dichloro-5-nitro-phenol, dichloronitrobenzenol, acmc-1afv8, ksc495i3b
InChI Key OFAPWTOOMSVMIU-UHFFFAOYSA-N
SMILES OC1=CC(=C(Cl)C=C1Cl)[N+]([O-])=O
IUPAC Name 2,4-dichloro-5-nitrophenol
Molecular Weight (g/mol) 207.99
PubChem CID 181635
Formula Weight 208
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-nitrophenol
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.