Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine

Catalog No. 7622961G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-229-61G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-229-61G Supplier Sigma Aldrich Supplier No. 7622961G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2,4-Dichloro-6,7-dihydro-5H-cyclopenta[d]pyrimidine

Specifications

CAS 5466-43-3
Melting Point 71°C to 79°C
Molecular Formula C7H6Cl2N2
Linear Formula C7H6Cl2N2
MDL Number MFCD09750204
Quantity 10 g
Synonym 2,4-Dichloro-5,6-trimethylenepyrimidine
Molecular Weight (g/mol) 189.04
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.