Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-6-nitrophenol, 98%, moist solid containing 20% water, Thermo Scientific™

Catalog No. AC209255000
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209255000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC209255000 Supplier Thermo Scientific Chemicals Supplier No. 209255000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-89-2 , 7732-18-5
Molecular Formula C6H3Cl2NO3
Molecular Weight (g/mol) 207.994
MDL Number MFCD00007101
InChI Key LYPMXMBQPXPNIQ-UHFFFAOYSA-N
Synonym 2,4-dichloro-6-nitrophenol, phenol, 2,4-dichloro-6-nitro, 2,4-dichlor-6-nitrofenol, 4,6-dichloro-2-nitrophenol, 2,4-dichlor-6-nitrofenol czech, pubchem22918, acmc-1aunw, wln: wnr bq cg eg, 2-nitro-4,6-dichlorophenol
PubChem CID 11871
SMILES C1=C(C=C(C(=C1Cl)O)[N+](=O)[O-])Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Cl2C6H2(NO2)OH
Quantity 500 g
Beilstein 06, 241
Formula Weight 208
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 2, 4-Dichloro-6-nitrophenol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.