Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine, 95%, Thermo Scientific™

Catalog No. AC454012500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454012500 250 mg
AC454010010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC454012500 Supplier Thermo Scientific Chemicals Supplier No. 454012500

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 90213-66-4
Molecular Formula C6H3Cl2N3
Molecular Weight (g/mol) 188.02
InChI Key GHXBPCSSQOKKGB-UHFFFAOYSA-N
Synonym 2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l
PubChem CID 14116871
IUPAC Name 2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine
SMILES C1=CNC2=C1C(=NC(=N2)Cl)Cl

Specifications

Melting Point 250°C to 252°C
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 188.02
Percent Purity 95%
Chemical Name or Material 2, 4-Dichloro-7H-pyrrolo[2, 3-d]pyrimidine
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.