Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloro-D-phenylalanine, 98%, Thermo Scientific™

Catalog No. AAH2726903
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2726903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2726903 Supplier Thermo Scientific Chemicals Supplier No. H2726903

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 114872-98-9
Molecular Formula C9H9Cl2NO2
Molecular Weight (g/mol) 234.076
MDL Number MFCD01860873
InChI Key GWHQTNKPTXDNRM-MRVPVSSYSA-N
Synonym 2,4-dichloro-d-phenylalanine, d-2,4-dichlorophenylalanine, 2r-2-amino-3-2,4-dichlorophenyl propanoic acid, r-2-amino-3-2,4-dichlorophenyl propanoic acid, d-2,4-dichlorophe, d-phenylalanine, 2,4-dichloro, h-d-phe 2,4-cl2-oh, 2r-2-amino-3-2,4-dichlorophenyl propionic acid, d-2,4-dichloro phenylalanine, 2 3h-furanone,dihydro-3,5-dimethyl-5-2-1-oxopropoxy ethyl
PubChem CID 6993648
IUPAC Name (2R)-2-amino-3-(2,4-dichlorophenyl)propanoic acid
SMILES C1=CC(=C(C=C1Cl)Cl)CC(C(=O)O)N

Specifications

Quantity 1 g
Formula Weight 234.09
Optical Rotation −31° (c=0.5 in 0.5M NaOH)
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-D-phenylalanine
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.