Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloroanisole, 99%

Catalog No. AAB2004222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2004222 100 g
AAB2004206 5 g
AAB2004214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2004222 Supplier Thermo Scientific Chemicals Supplier No. B2004222
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 553-82-2 | C7H6Cl2O | 177.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-82-2
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00044772
InChI Key CICQUFBZCADHHX-UHFFFAOYSA-N
Synonym 2,4-dichloroanisole, benzene, 2,4-dichloro-1-methoxy, 1,5-dichloro-2-methoxybenzene, 2,4-dichloro-1-methoxy-benzene, anisole, 2,4-dichloro, 2,4-dichloromethoxybenzene, anisole,4-dichloro, acmc-209lmo, ksc273i7j
PubChem CID 11119
IUPAC Name 2,4-dichloro-1-methoxybenzene
SMILES COC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point 24°C to 27°C
Density 1.288
Boiling Point 125°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5615
Quantity 100 g
Beilstein 1940358
Formula Weight 177.03
Percent Purity 99%
Chemical Name or Material 2,4-Dichloroanisole
Hazard Category H302+H332-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 209-051-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.