Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloroanisole 97.0+%, TCI America™
SDP

Catalog No. D032625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D032625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D032625G Supplier TCI America Supplier No. D032625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 553-82-2
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
MDL Number MFCD00044772
InChI Key CICQUFBZCADHHX-UHFFFAOYSA-N
Synonym 2,4-dichloroanisole, benzene, 2,4-dichloro-1-methoxy, 1,5-dichloro-2-methoxybenzene, 2,4-dichloro-1-methoxy-benzene, anisole, 2,4-dichloro, 2,4-dichloromethoxybenzene, anisole,4-dichloro, acmc-209lmo, ksc273i7j
PubChem CID 11119
IUPAC Name 2,4-dichloro-1-methoxybenzene
SMILES COC1=CC=C(Cl)C=C1Cl

Specifications

Color White
Quantity 25 g
Formula Weight 177.02
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichloroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.