Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D40375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D40375G 5 g
D403725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D40375G Supplier TCI America Supplier No. D40375G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16271-33-3
Molecular Formula C6H3Cl3O2S
Molecular Weight (g/mol) 245.498
MDL Number MFCD00052712
InChI Key FDTPBIKNYWQLAE-UHFFFAOYSA-N
Synonym 2,4-dichloro-benzenesulfonyl chloride, 2,4-dichlorobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 2,4-dichloro, 2,4-dichlorobenzenesulphonyl chloride, 2,4-dichlorobenzenesulfonylchloride, 2,4-dichlorophenylsulfonyl chloride, 2,4-dichlorobenzenesulphonylchloride, 2,4-dichlorophenyl chlorosulfone, pubchem5115, acmc-1c9b8
PubChem CID 204419
IUPAC Name 2,4-dichlorobenzenesulfonyl chloride
SMILES C1=CC(=C(C=C1Cl)Cl)S(=O)(=O)Cl

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 5 g
UN Number 3261
Formula Weight 245.50
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Dichlorobenzenesulfonyl Chloride
RTECSNumber DB8927500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.