Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dichlorobenzophenone 99.0+%, TCI America™
SDP

Catalog No. D023025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D023025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D023025G Supplier TCI America Supplier No. D023025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 85-29-0
Molecular Formula C13H8Cl2O
Molecular Weight (g/mol) 251.11
MDL Number MFCD00038744
InChI Key YXMYPHLWXBXNFF-UHFFFAOYSA-N
Synonym 2,4'-dichlorobenzophenone, 2-chlorophenyl 4-chlorophenyl methanone, methanone, 2-chlorophenyl 4-chlorophenyl, 2-chlorophenyl-4-chlorophenyl methanone, unii-h3n0v78i65, benzophenone, 2,4'-dichloro, di2-chlorophenyl ketone, pubchem3373, o,p-dichlorobenzophenone, o,p'-dichlorobenzophenone
PubChem CID 66558
IUPAC Name (2-chlorophenyl)(4-chlorophenyl)methanone
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=CC=C1Cl

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 214°C
Quantity 25 g
Formula Weight 251.11
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4′-Dichlorobenzophenone
EINECSNumber (4)-1353
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.