Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzoyl chloride, 98%

Catalog No. AC150221000
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150221000 100 mL Glass bottle
AC150220050 5 mL Glass bottle
AC150225000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC150221000 Supplier Thermo Scientific Chemicals Supplier No. 150221000
Only null left
Add to Cart

CAS: 89-75-8 | C7H3Cl3O | 209.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-75-8
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.45
MDL Number MFCD00000661
InChI Key CEOCVKWBUWKBKA-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,4-dichloro, 2,4-dichlorobenzoylchloride, benzoylchloride, 2,4-dichloro, ccris 8616, acmc-1cuf7, dsstox_cid_31176, dsstox_gsid_52603, 2,4-dichloro benzoylchloride, 2.4-dichlorobenzoyl chloride, ksc450c2p
PubChem CID 66645
IUPAC Name 2,4-dichlorobenzoyl chloride
SMILES ClC(=O)C1=CC=C(Cl)C=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 21°C
Color Colorless
Density 1.4900g/mL
Boiling Point 150°C (34.0 mmHg)
Flash Point 137°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5915 to 1.5937
Linear Formula Cl2C6H3COCl
Quantity 100 mL
Beilstein 09, 342
Specific Gravity 1.49
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in alcohol,ether and acetone,slightly soluble in heptane
Formula Weight 209.46
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material 2, 4-Dichlorobenzoyl chloride, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 201-936-4
RTECSNumber DM6636733
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.