Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzoyl Chloride 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
Packaging:
Glass bottle
89-75-8
C7H3Cl3O
209.46
MFCD00000661
CEOCVKWBUWKBKA-UHFFFAOYSA-N
benzoyl chloride, 2,4-dichloro,2,4-dichlorobenzoylchloride,benzoylchloride, 2,4-dichloro,ccris 8616,acmc-1cuf7,dsstox_cid_31176,dsstox_gsid_52603,2,4-dichloro benzoylchloride,2.4-dichlorobenzoyl chloride,ksc450c2p
66645
2,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1Cl)Cl)C(=O)Cl
89-75-8
C7H3Cl3O
209.46
MFCD00000661
CEOCVKWBUWKBKA-UHFFFAOYSA-N
benzoyl chloride, 2,4-dichloro,2,4-dichlorobenzoylchloride,benzoylchloride, 2,4-dichloro,ccris 8616,acmc-1cuf7,dsstox_cid_31176,dsstox_gsid_52603,2,4-dichloro benzoylchloride,2.4-dichlorobenzoyl chloride,ksc450c2p
66645
2,4-dichlorobenzoyl chloride
C1=CC(=C(C=C1Cl)Cl)C(=O)Cl
Refractive Index 1.5915 to 1.5937
Density 1.4900g/mL
Melting Point 21°C
Boiling Point 150°C (34.0mmHg)
Flash Point 137°C
Specific Gravity 1.49
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 500mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula Cl2C6H3COCl
Beilstein 09, 342
Solubility Information Solubility in water: may decompose. Other solubilities: soluble in alcohol,ether and acetone,slightly soluble in heptane
Formula Weight 209.46
Physical Form Liquid After Melting
Percent Purity ≥97.5% (RCOCL)
Chemical Name or Material 2, 4-Dichlorobenzoyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-936-4

RTECSNumber : DM6636733

TSCA : TSCA