Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorobenzyl Alcohol, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18620659 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
25 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-659 25 g
18-620-660 100 g
18-620-661 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-620-659 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D285625GM
Add to Cart
Add to Cart

Synonym: (2,4-Dichlorophenyl)methanol, Dybenal, 2,4-Dichlorobenzenemethanol

Chemical Identifiers

CAS 1777-82-8
Molecular Formula C7H6Cl2O
Molecular Weight (g/mol) 177.02
InChI Key DBHODFSFBXJZNY-UHFFFAOYSA-N
IUPAC Name (2,4-dichlorophenyl)methanol
SMILES OCC1=CC=C(Cl)C=C1Cl

Specifications

Quantity 25 g
Grade Reagent
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.