Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4'-Dichlorobiphenyl solution, 100μg/mL in hexane, Ultra Scientific

Catalog No. USRPC089S
Encompass
Change view
Click to view available options
Quantity:
1 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-RPC-089S 1 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. US-RPC-089S Supplier Agilent Technologies Supplier No. RPC089S
May include imposed supplier surcharges.
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 34883-43-7 , 110-54-3
Molecular Formula C12H8Cl2
Molecular Weight (g/mol) 223.096
InChI Key UFNIBRDIUNVOMX-UHFFFAOYSA-N
PubChem CID 36982
ChEBI CHEBI:34232
IUPAC Name 1-chloro-2-(4-chlorophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=C(C=C2)Cl)Cl

Specifications

CAS Max % 99.98, 0.02
Analyte or Component Names n-hexane,2,4'-dichlorobiphenyl
Element n-hexane, 2,4'-dichlorobiphenyl
Solvent or Matrix Hexane
Quantity 1 x 2 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material 2, 4'-Dichlorobiphenyl solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.