Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenol, 99%, Thermo Scientific™

Catalog No. AAA1206536
Change view
Click to view available options
Quantity:
500 g
2500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1206536 500 g
AAA120650E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AAA1206536 Supplier Thermo Scientific Chemicals Supplier No. A1206536

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-83-2
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002169
InChI Key HFZWRUODUSTPEG-UHFFFAOYSA-N
Synonym 4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
PubChem CID 8449
ChEBI CHEBI:16738
IUPAC Name 2,4-dichlorophenol
SMILES OC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point 40°C to 46°C
Boiling Point 209°C to 210°C
Flash Point 114°C (237°F)
Odor Phenol-like
Quantity 500 g
UN Number UN2928
Beilstein 742467
Merck Index 14,3072
Formula Weight 163
Percent Purity 99%
Chemical Name or Material 2,4-Dichlorophenol
Show More Show Less
Hazard Category H302-H311-H314-H331-H335-H350-H500
Hazard Statement GHS H Statement
H311-H314-H318-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
EINECSNumber 204-429-6
RTECSNumber SK8575000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.