Learn More
Research Products International Corp 2,4-Dichlorophenoxyacetic Acid [2,4 D], 250 Grams
SDP
Supplier:  Research Products International Corp D43050250.0
Encompass_Preferred
Synthetic auxin like growth regulator and as such it is often used in laboratories for plant research and as a supplement in plant cell culture media such as MS medium.
Catalog No. 50-213-288