Learn More
Bioworld 2,4-Dichlorophenoxyacetic Acid, 100 g
Supplier:  Bioworld 306310011
Encompass_Preferred
2,4-Dichlorophenoxyacetic acid2,4-D is one of the most extensively researched and studied crop protection tool used in agriculture. The product is authorized for use in almost 70 countries worldwide, including the United States, Canada, the United Kingdom, Australia, New Zealand, Germany, France and Japan. 2,4-D does not present an unacceptable risk to human health or the environment when used properly. bio-world supplies this product for research use only, used in tissue culture as a plant growth regulator.Auxin like growth regulatorPlant tissue culture tested
Catalog No. 50-255-031