Learn More
Medchemexpress LLC 2,4-dichlorophenoxyacetic acid | 94-75-7 | 99.9% | 221.04 | 250 MG
SDP
Catalog No. 5000385457
Encompass_Preferred
2,4-D (Standard) is an analytical standard intended for research and analytical applications. Also known as 2,4-dichlorophenoxyacetic acid, it acts as a selective herbicide for broad-leaved weed control. This compound induces apoptosis and inhibits DNA and protein synthesis, preventing normal plant growth and development.
  • Analytical standard for research and analytical applications
  • Selective herbicide for broad-leaved weed control
  • Induces apoptosis and inhibits DNA and protein synthesis
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-85457 Supplier Medchemexpress LLC Supplier No. 250MGHY18572R
2,4-D (Standard) is an analytical standard intended for research and analytical applications. Also known as 2,4-dichlorophenoxyacetic acid, it acts as a selective herbicide for broad-leaved weed control. This compound induces apoptosis and inhibits DNA and protein synthesis, preventing normal plant growth and development.
  • Analytical standard for research and analytical applications
  • Selective herbicide for broad-leaved weed control
  • Induces apoptosis and inhibits DNA and protein synthesis
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC, and MS