Learn More
Medchemexpress LLC 2,4-Dichlorophenoxyacetic acid sodium | 2702-72-9 | ≥98.0% | 243.02 | 25 MG
SDP
Catalog No. 5000384045
Encompass_Preferred
2,4-D sodium is a selective herbicide used for the control of broad-leaved weeds. It can induce apoptosis and inhibits DNA and protein synthesis, thereby preventing normal plant growth and development.
  • Selective herbicide for broad-leaved weeds.
  • Induces apoptosis.
  • Inhibits DNA and protein synthesis.
Catalog No. 50-003-84045 Supplier Medchemexpress LLC Supplier No. 25MGHY18572AR
2,4-D sodium is a selective herbicide used for the control of broad-leaved weeds. It can induce apoptosis and inhibits DNA and protein synthesis, thereby preventing normal plant growth and development.
  • Selective herbicide for broad-leaved weeds.
  • Induces apoptosis.
  • Inhibits DNA and protein synthesis.