Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichlorophenylacetone 98.0+%, TCI America™
SDP

Catalog No. D29285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29285G 5 g
D292825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29285G Supplier TCI America Supplier No. D29285G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 93457-07-9
Molecular Formula C9H8Cl2O
Molecular Weight (g/mol) 203.06
MDL Number MFCD00210405
InChI Key ITVVHXZQWSCZBC-UHFFFAOYSA-N
Synonym 2,4-dichlorophenylacetone, 1-2,4-dichlorophenyl propan-2-one, 1-2,4-dichlorophenyl-2-propanone, 1-2,4-dichlorophenyl acetone, acmc-209rlw, ghl.pd_mitscher_leg0.657, 2,4-dichlorophenyl-2-propanone, 1-2,4-dichlorophenyl-propan-2-one
PubChem CID 2734102
IUPAC Name 1-(2,4-dichlorophenyl)propan-2-one
SMILES CC(=O)CC1=CC=C(Cl)C=C1Cl

Specifications

Melting Point 49°C
Color White-Yellow
Quantity 5 g
Formula Weight 203.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,4-Dichlorophenylacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.