Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. D37845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37845G 5 g
D378425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37845G Supplier TCI America Supplier No. D37845G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 26452-80-2 | C5H3Cl2N | 147.986 g/mol

Chemical Identifiers

CAS 26452-80-2
Molecular Formula C5H3Cl2N
Molecular Weight (g/mol) 147.986
MDL Number MFCD00955618
InChI Key TYPVHTOETJVYIV-UHFFFAOYSA-N
Synonym pyridine, 2,4-dichloro, 2,4-dichloro pyridine, 2,4-dichoropyridine, 2, 4-dichloropyridine, 2,4-dichloro-pyridine, 2,4-dichlorpyridin, 2,4-dichlorpyridine, 2,4-dichloropyrdine, pubchem3531, acmc-1cnvt
PubChem CID 33515
IUPAC Name 2,4-dichloropyridine
SMILES C1=CN=C(C=C1Cl)Cl

Specifications

Boiling Point 190°C
Quantity 5 g
Formula Weight 147.99
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Dichloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.