Learn More
AdipoGen 2 4-Dichlororesorcinol
SDP
Catalog No. 502561108
Encompass_Preferred
Chemical. CAS 16606-61-4. Formula C6H4Cl2O2. MW 179. Synthetic. Bulding block for synthesis. Reactant used for the preparation of fluorescent compounds.
Catalog No. 50-256-1108 Supplier AdipoGen Supplier No. CDXD0334G010
Chemical. CAS 16606-61-4. Formula C6H4Cl2O2. MW 179. Synthetic. Bulding block for synthesis. Reactant used for the preparation of fluorescent compounds.