Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 5g, 2,4-Dichlorothieno[3,2-d]pyrimidine, >95%, [16234-14-3], D14603 205.064 C6H2Cl2N2S

Catalog No. 501409000
Encompass_Preferred

Compound: 2,4-Dichlorothieno[3,2-d]pyrimidine

CAS#: 16234-14-3

MDL#: not listed

Background: A dichlorinated thienopyrimidine used as a building block in pharmaceutical research. The two chlorine substituents allow for sequential, regioselective cross-coupling or nucleophilic substitution reactions on this fused bicyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-140-9000 Supplier Frontier Specialty Chemicals Supplier No. D146035G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2,4-Dichlorothieno[3,2-d]pyrimidine

CAS#: 16234-14-3

MDL#: not listed

Background: A dichlorinated thienopyrimidine used as a building block in pharmaceutical research. The two chlorine substituents allow for sequential, regioselective cross-coupling or nucleophilic substitution reactions on this fused bicyclic scaffold.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.