Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dichlorothieno[3,2-d]pyrimidine

Catalog No. CDS005155 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS005155 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS005155 Supplier Sigma Aldrich Supplier No. CDS005155100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The postfunctionalization reaction of 2-thiophenecarbonyl chloride with single walled carbon nanotubes (SWCNTs) was studied.2-Thiophenecarbonyl chloride was used in the synthesis of building blocks derived from ornithine by undergoing acylation/sulphonation of copper complex of orthinine.

Specifications

Molecular Formula C6H2Cl2N2S
Linear Formula C6H2Cl2N2S
Quantity 100 mg
MDL Number MFCD08448158
Molecular Weight (g/mol) 205.06
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.