Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,4-Dichlorotoluene

Catalog No. 1450091KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-500-91KG 1 kg
14-500-9250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-500-91KG Supplier Sigma Aldrich Supplier No. 1450091KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 2,4-dichlorotoluene was used as an oxidant and a solvent during anaerobic catalytic oxidation of secondary alcohols by using an in situ (n-heterocyclic carbene)-ni(0) system
  • It was used as growth supplement in the culture media of ralstonia sp
  • Strain ps12
  • It was used to develop a sensitive method based on solid-phase microextraction followed by gas chromatography-tandem mass spectrometry for the analysis of chlorotoluenes in water samples

Specifications

CAS 95-73-8
Density 1.246 g/mL (at 25°C (literature))
Boiling Point 200°C (lit.)
Molecular Formula C7H6Cl2
Refractive Index n20/D 1.546 (literature)
Linear Formula CH3C6H3Cl2
MDL Number MFCD00000583
Quantity 1 kg
Molecular Weight (g/mol) 161.03
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.