Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Diethyl-2,6-heptadienal, 97%, mixture of isomers, Thermo Scientific™

Catalog No. AC218935000
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC218935000 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC218935000 Supplier Thermo Scientific Chemicals Supplier No. 218935000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 85136-07-8
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C11H18O
Refractive Index 1.4650 to 1.4750 (20°C, 589nm)
Linear Formula H2C=CHCH2CH(C2H5)CH=C(C2H5)CHO
Quantity 500g
Synonym 2e-2,4-diethylhepta-2,6-dienal, 2,4-diethyl-2,6-heptadienal, e-2,4-diethylhepta-2,6-dienal, 2e-2,4-diethyl-2,6-heptadienal
InChI Key QAFGEOHWEQLURJ-FLIBITNWSA-N
SMILES CCC(CC=C)C=C(CC)C=O
IUPAC Name (2Z)-2,4-diethylhepta-2,6-dienal
Molecular Weight (g/mol) 166.264
PubChem CID 16211670
Formula Weight 166.26
Percent Purity ≥96.0%
Chemical Name or Material 2, 4-Diethyl-2, 6-heptadienal, 97%
Show More Show Less
EINECSNumber 285-729-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.