Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Difluoroacetophenone, 98%

Catalog No. AAA1342014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1342014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1342014 Supplier Thermo Scientific Chemicals Supplier No. A1342014

missing translation for 'validMainframeMsg'

CAS: 364-83-0 | C8H6F2O | 156.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 364-83-0
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD00151261
InChI Key QEWHNJPLPZOEKU-UHFFFAOYSA-N
Synonym 2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213
PubChem CID 67770
IUPAC Name 1-(2,4-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=C(C=C1)F)F

Specifications

Density 1.232
Boiling Point 80°C to 81°C (25 mmHg)
Flash Point 66°C (150°F)
Odor Characteristic
Refractive Index 1.488
Quantity 25 g
Beilstein 1940741
Formula Weight 156.13
Percent Purity 98%
Chemical Name or Material 2',4'-Difluoroacetophenone
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 206-667-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.