Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Difluoroanisole 99.0+%, TCI America™
SDP

Catalog No. D184910G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D184910G 10 g
D184925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D184910G Supplier TCI America Supplier No. D184910G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 452-10-8
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.12
MDL Number MFCD00010731
InChI Key CRMJLJFDPNJIQA-UHFFFAOYSA-N
Synonym 2,4-difluoroanisole, 2,4-difluoroanilsole, 2,4-difluoromethoxy benzene, benzene, 2,4-difluoro-1-methoxy, pubchem3455, 2,4-difluoro anisole, ksc493m8b, 2,4-difluoro-1-methoxy-benzene, #
PubChem CID 136293
IUPAC Name 2,4-difluoro-1-methoxybenzene
SMILES COC1=CC=C(F)C=C1F

Specifications

Color Yellow
Boiling Point 53°C
Quantity 10 g
UN Number 1993
Formula Weight 144.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluoroanisole
EINECSNumber (3)-4291
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.