Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,4-Difluorophenyl)-4-piperidylmethanone Oxime Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D42695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42695G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D42695G Supplier TCI America Supplier No. D42695G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 135634-18-3
Molecular Formula C12H15ClF2N2O
Molecular Weight (g/mol) 276.712
MDL Number MFCD11869788
InChI Key CPVWKXXFKMUDPA-UHFFFAOYSA-N
PubChem CID 57357481
IUPAC Name N-[(2,4-difluorophenyl)-piperidin-4-ylmethylidene]hydroxylamine;hydrochloride
SMILES C1CNCCC1C(=NO)C2=C(C=C(C=C2)F)F.Cl

Specifications

Color White
Quantity 5 g
Formula Weight 276.71
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (2,4-Difluorophenyl)-4-piperidylmethanone Oxime Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.