Learn More
Medchemexpress LLC 2,4-Difluorophenylboronic acid | 144025-03-6 | 99.93% | 157.91 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY32883
Encompass_Preferred
2,4-Difluorophenylboronic acid is an intermediate that can be used to synthesize dual FGFR and VEGFR inhibitors. It can also be used in the research of angiogenesis and cancer.
  • Intermediate for synthesis.
  • Can be used to synthesize dual FGFR and VEGFR inhibitors.
  • Applicable in angiogenesis research.
  • Applicable in cancer research.
Catalog No. 50-004-03014