Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',4'-Difluoropropiophenone, 98%, Thermo Scientific™

Catalog No. AC290680100
Change view
Click to view available options
Quantity:
10 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC290680100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC290680100 Supplier Thermo Scientific Chemicals Supplier No. 290680100

Chemical Identifiers

CAS 85068-30-0
Molecular Formula C9H8F2O
Molecular Weight (g/mol) 170.16
MDL Number MFCD00015507
InChI Key UZWOADNMVRRYDE-UHFFFAOYSA-N
Synonym 2',4'-difluoropropiophenone, 1-2,4-difluorophenyl propan-1-one, 2,4-difluoropropiophenone, 1-propanone, 1-2,4-difluorophenyl, 2,4-defluoropropiophenone, 1-2,4-difluorophenyl-1-propanone, 1-2.4-difluoro-phenyl-propan-1-one, acmc-209q3o, ksc654a2r
PubChem CID 123586
IUPAC Name 1-(2,4-difluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(F)C=C1F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1930g/mL
Boiling Point 51.0°C to 52.0°C (1.5 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 10 g
Specific Gravity 1.193
Solubility Information Solubility in water: insoluble.,soluble in most common organic solvents
Formula Weight 170.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2', 4'-Difluoropropiophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 285-291-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.