Learn More
Medchemexpress LLC 2,4-difluororesorcinol | 195136-71-1 | MFCD09836234 | 99.6% | 146.09 g·mol⁻1 | C6H4F2O2 | 10MM 1ML
SDP
Catalog No. 5000239295
Encompass_Preferred
2,4-Difluororesorcinol is a fluorinated resorcinol building block used in organic and medicinal chemistry as a precursor for fluorescent dyes and synthetic intermediates. It is offered as a solid or as a ready-to-use 10 mM solution in DMSO and is characterized by a molecular weight of 146.09 g·mol⁻1 and CAS 195136-71-1.
  • Fluorinated resorcinol building block for chemical synthesis.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • High purity (about 99.6%).
  • Molecular weight 146.09 g·mol⁻1 and CAS 195136-71-1 for unambiguous identification.
  • Brown to reddish brown solid appearance.
  • Recommended cold storage for long-term stability (powder at -20°C; solution at -80°C).
Catalog No. 50-002-39295 Supplier Medchemexpress LLC Supplier No. HY12447210MM 1ML
Add to Cart
Add to Cart
2,4-Difluororesorcinol is a fluorinated resorcinol building block used in organic and medicinal chemistry as a precursor for fluorescent dyes and synthetic intermediates. It is offered as a solid or as a ready-to-use 10 mM solution in DMSO and is characterized by a molecular weight of 146.09 g·mol⁻1 and CAS 195136-71-1.
  • Fluorinated resorcinol building block for chemical synthesis.
  • Supplied as solid and as a 10 mM solution in DMSO for convenience.
  • High purity (about 99.6%).
  • Molecular weight 146.09 g·mol⁻1 and CAS 195136-71-1 for unambiguous identification.
  • Brown to reddish brown solid appearance.
  • Recommended cold storage for long-term stability (powder at -20°C; solution at -80°C).